4-Hydroxy-3-methylphenyl thiocyanate ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Methyl-4-thiocyanatophenol
043354-500MG
043354-1G
043354-5G
101860-398EA
90.77
USD
101860-398
101860-400
101860-402
4-Hydroxy-3-methylphenyl thiocyanate ≥95%
4-Hydroxy-3-methylphenyl thiocyanate
Hazard: Irritant
Formula:
C₈H₇NOS MW: 165.22 g/mol Melting Pt: 73…75 °C |
MDL Number:
MFCD00086627 CAS Number: 3774-53-6 |
Specification Test Results
Melting point | 73-75°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...