2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Bromo-1-(5-chlorothiophen-2-yl)ethanone, 2-(Bromoacetyl)-5-chlorothiophene
043353-500MG
043353-1G
043353-5G
101860-392EA
278.46
USD
101860-392
101860-394
101860-396
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone ≥95%
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone
Hazard: Irritant
Formula:
C₆H₄BrClOS MW: 239.52 g/mol Melting Pt: 73…75 °C |
MDL Number:
MFCD00829820 CAS Number: 57731-17-6 |
Specification Test Results
Melting point | 73-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...