3-(4-Fluorobenzyl)-5,5-dimethyl-1H-imidazole-2,4(3H,5H)-dione ≥95%
Supplier: Matrix Scientific
043352-500MG
043352-1G
043352-5G
101860-386EA
181.54
USD
101860-386
101860-388
101860-390
3-(4-Fluorobenzyl)-5,5-dimethyl-1H-imidazole-2,4(3H,5H)-dione ≥95%
3-(4-Fluorobenzyl)-5,5-dimethyl-1H-imidazole-2,4(3H,5H)-dione
Hazard: Irritant
Formula:
C₁₂H₁₃FN₂O₂ MW: 236.25 g/mol Melting Pt: 94…95 °C |
MDL Number:
MFCD05975052 CAS Number: 860787-36-6 |
Specification Test Results
Melting point | 94-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...