Ethyl-5-(4-methoxyphenyl)-2-thiophenecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 5-(4-methoxyphenyl)thiophene-2-carboxylate
043348-500MG
043348-1G
043348-5G
101860-370EA
747.69
USD
101860-370
101860-372
101860-374
Ethyl-5-(4-methoxyphenyl)-2-thiophenecarboxylate ≥95%
Ethyl-5-(4-methoxyphenyl)-2-thiophenecarboxylate
Hazard: Irritant
Formula:
C₁₄H₁₄O₃S MW: 262.33 g/mol Melting Pt: 71…75 °C |
MDL Number:
MFCD01860770 CAS Number: 13858-71-4 |
Specification Test Results
Melting point | 71-75°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...