Pentafluorophenyl 5-chloro-2-methoxybenzenesulfonate ≥95%
Supplier: Matrix Scientific
043346-500MG
043346-1G
043346-5G
101860-364EA
375.38
USD
101860-364
101860-366
101860-368
Pentafluorophenyl 5-chloro-2-methoxybenzenesulfonate ≥95%
Pentafluorophenyl 5-chloro-2-methoxybenzenesulphonate
Hazard: Irritant
Formula:
C₁₃H₆ClF₅O₄S Melting Pt: 129-131°C |
MDL Number:
MFCD05975152 CAS Number: 886361-33-7 |
Specification Test Results
Melting point | 129-131°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...