3-[(2,3,4,5,6-Pentafluorophenoxy)sulfonyl]benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-[(2,3,4,5,6-Pentafluorophenoxy)sulphonyl]benzoic acid

043345-500MG 043345-1G 043345-5G
101860-358EA 375.38 USD
101860-358 101860-360 101860-362
3-[(2,3,4,5,6-Pentafluorophenoxy)sulfonyl]benzenecarboxylic acid ≥95%
3-[(2,3,4,5,6-Pentafluorophenoxy)sulfonyl]benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₅F₅O₅S
MW: 368.24 g/mol
Melting Pt: 130…132 °C
MDL Number: MFCD05975151
CAS Number: 886361-31-5

Order Now

Specification Test Results

Melting point 130-132°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR