Pentafluorophenyl 4-(2-nitro-4-(trifluoromethyl)phenoxy)benzenesulfonate ≥95%

Supplier: Matrix Scientific

043344-500MG 043344-1G 043344-5G
101860-352EA 375.38 USD
101860-352 101860-354 101860-356
Pentafluorophenyl 4-(2-nitro-4-(trifluoromethyl)phenoxy)benzenesulfonate ≥95%
Pentafluorophenyl 4-(2-nitro-4-(trifluoromethyl)phenoxy)benzenesulphonate

Hazard: Irritant

Formula: C₁₉H₇F₈NO₆S
Melting Pt: 107…109 °C
MDL Number: MFCD05975150
CAS Number: 886361-29-1

Order Now

Specification Test Results

Melting point 107-109°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR