Pentafluorophenyl 4-bromo-2-chlorobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043341-500MG 043341-1G 043341-5G
101860-334EA 375.38 USD
101860-334 101860-336 101860-338
Pentafluorophenyl 4-bromo-2-chlorobenzenesulfonate ≥95%
Pentafluorophenyl 4-bromo-2-chlorobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₃BrClF₅O₃S
Melting Pt: 88…89 °C
MDL Number: MFCD05975147
CAS Number: 886361-22-4

Order Now

Specification Test Results

Melting point 88-89°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR