Pentafluorophenyl 2,4,6-triisopropylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

043340-500MG 043340-1G 043340-5G
101860-328EA 375.38 USD
101860-328 101860-330 101860-332
Pentafluorophenyl 2,4,6-triisopropylbenzenesulfonate ≥95%
Pentafluorophenyl 2,4,6-triisopropylbenzenesulphonate

Hazard: Irritant

Formula: C₂₁H₂₃F₅O₃S
Melting Pt: 42-43°C
MDL Number: MFCD05975146
CAS Number: 886361-20-2

Order Now

Specification Test Results

Melting point 42-43°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR