Pentafluorophenyl 1-naphthalenesulfonate ≥95%

Supplier: Matrix Scientific

043339-500MG 043339-1G 043339-5G
101860-322EA 375.38 USD
101860-322 101860-324 101860-326
Pentafluorophenyl 1-naphthalenesulfonate ≥95%
Pentafluorophenyl 1-naphthalenesulphonate

Hazard: Irritant

Formula: C₁₆H₇F₅O₃S
Melting Pt: 102-104°C
MDL Number: MFCD05975145
CAS Number: 886361-18-8

Order Now

Specification Test Results

Melting point 102-104°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR