Pentafluorophenyl pentamethylbenzenesulfonate ≥95%

Supplier: Matrix Scientific

043338-500MG 043338-1G 043338-5G
101860-316EA 375.38 USD
101860-316 101860-318 101860-320
Pentafluorophenyl pentamethylbenzenesulfonate ≥95%
Pentafluorophenyl pentamethylbenzenesulphonate

Hazard: Irritant

Formula: C₁₇H₁₅F₅O₃S
Melting Pt: 151…153 °C
MDL Number: MFCD05975144
CAS Number: 886361-16-6

Order Now

Specification Test Results

Melting point 151-153°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR