Methyl-2-[(2,3,4,5,6-pentafluorophenoxy)sulfonyl]benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043337-500MG 043337-1G 043337-5G
101860-310EA 260 USD
101860-310 101860-312 101860-314
Methyl-2-[(2,3,4,5,6-pentafluorophenoxy)sulfonyl]benzenecarboxylate ≥95%
Methyl-2-[(2,3,4,5,6-pentafluorophenoxy)sulfonyl]benzenecarboxylate

Hazard: Irritant

Formula: C₁₄H₇F₅O₅S
MW: 382.26 g/mol
Melting Pt: 59…61 °C
MDL Number: MFCD05975143
CAS Number: 886361-14-4

Order Now

Specification Test Results

Melting point 59-61°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR