Pentafluorophenyl 4-bromobenzenesulfonate ≥95%

Supplier: Matrix Scientific

043336-500MG 043336-1G 043336-5G
101860-304EA 260 USD
101860-304 101860-306 101860-308
Pentafluorophenyl 4-bromobenzenesulfonate ≥95%
Pentafluorophenyl 4-bromobenzenesulphonate

Hazard: Irritant

Formula: C₁₂H₄BrF₅O₃S
Melting Pt: 66…68 °C
MDL Number: MFCD00404028
CAS Number: 848649-38-7

Order Now

Specification Test Results

Melting point 66-68°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR