Pentafluorophenyl 3,4-dimethoxybenzenesulfonate ≥95%

Supplier: Matrix Scientific

043335-500MG 043335-1G 043335-5G
101860-298EA 375.38 USD
101860-298 101860-300 101860-302
Pentafluorophenyl 3,4-dimethoxybenzenesulfonate ≥95%
Pentafluorophenyl 3,4-dimethoxybenzenesulphonate

Hazard: Irritant

Formula: C₁₄H₉F₅O₅S
Melting Pt: 93…95 °C
MDL Number: MFCD05975142
CAS Number: 886361-12-2

Order Now

Specification Test Results

Melting point 93-95°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR