1-[(6-Chloro-3-pyridinyl)methyl]-4-phenylpiperazine ≥95%

Supplier: Matrix Scientific

043333-500MG 043333-1G 043333-5G
101860-286EA 278.46 USD
101860-286 101860-288 101860-290
1-[(6-Chloro-3-pyridinyl)methyl]-4-phenylpiperazine ≥95%
1-[(6-Chloro-3-pyridinyl)methyl]-4-phenylpiperazine

Hazard: Irritant

Formula: C₁₆H₁₈ClN₃
Melting Pt: 133…136 °C
MDL Number: MFCD04124602
CAS Number: 860787-10-6

Order Now

Specification Test Results

Melting point 133-136°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR