2-Chloronicotinaldehyde oxime ≥95%
Supplier: Matrix Scientific
043331-500MG
043331-1G
043331-5G
101860-280EA
556.92
USD
101860-280
101860-282
101860-284
2-Chloronicotinaldehyde oxime ≥95%
2-Chloronicotinaldehyde oxime
Hazard: Irritant
Formula:
C₆H₅ClN₂O Melting Pt: 165…167 °C |
MDL Number:
MFCD04117788 CAS Number: 215872-96-1 |
Specification Test Results
Melting point | 165-167°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...