4-(2-Chloroacetyl)-1H-pyrrole-2-carbaldehyde ≥95%
Supplier: Matrix Scientific
043329-500MG
043329-1G
043329-5G
101860-270EA
118.46
USD
101860-270
101860-272
101860-274
4-(2-Chloroacetyl)-1H-pyrrole-2-carbaldehyde ≥95%
4-(2-Chloroacetyl)-1H-pyrrole-2-carbaldehyde
Hazard: Irritant
Formula:
C₇H₆ClNO₂ Melting Pt: 179…181 °C |
MDL Number:
MFCD04973717 CAS Number: 115027-23-1 |
Specification Test Results
Melting point | 179-181°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...