Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3,4-dihydro-2-methyl-3-oxo-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylate
043327-500MG
043327-5G
101860-258EA
278.46
USD
101860-258
101860-262
Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylate ≥95%
Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-2-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₂N₂O₄ MW: 236.23 g/mol Melting Pt: 143…145 °C |
MDL Number:
MFCD04124547 CAS Number: 154365-38-5 |
Specification Test Results
Melting point | 143-145°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...