2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

043326-500MG 043326-1G 043326-5G
101860-252EA 169.23 USD
101860-252 101860-254 101860-256
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid ≥95%
2-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₄
MW: 207.19 g/mol
Melting Pt: 192…194 °C
MDL Number: MFCD04124544
CAS Number: 154365-40-9

Order Now

Specification Test Results

Melting point 192-194°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR