Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Supplier: Matrix Scientific
043325-500MG
043325-1G
043325-5G
101860-246EA
278.46
USD
101860-246
101860-248
101860-250
Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate ≥95%
Ethyl-2-methyl-3-oxo-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₃NO₄ MW: 235.24 g/mol Melting Pt: 141…143 °C |
MDL Number:
MFCD04124540 CAS Number: 154365-33-0 |
Specification Test Results
Melting point | 141-143°C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...