Dimethyl-2-[2-nitro-4-(piperidinosulfonyl)phenyl]malonate ≥95%

Supplier: Matrix Scientific

043322-500MG 043322-1G 043322-5G
101860-228EA 556.92 USD
101860-228 101860-230 101860-232
Dimethyl-2-[2-nitro-4-(piperidinosulfonyl)phenyl]malonate ≥95%
Dimethyl-2-[2-nitro-4-(piperidinosulfonyl)phenyl]malonate

Hazard: Irritant

Formula: C₁₆H₂₀N₂O₈S
Melting Pt: 109…111 °C
MDL Number: MFCD04124523
CAS Number: 860785-48-4

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR