N-(2-Chlorophenyl)-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

043319-500MG 043319-1G 043319-5G
101860-210EA 278.46 USD
101860-210 101860-212 101860-214
N-(2-Chlorophenyl)-1-hydrazinecarboxamide ≥95%
N-(2-Chlorophenyl)-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₇H₈ClN₃O
Melting Pt: 114…116 °C
MDL Number: MFCD03791165
CAS Number: 35580-76-8

Order Now

Specification Test Results

Melting point 114-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR