3-(2-Chlorophenyl)-1H-pyrazole ≥95%

Supplier: Matrix Scientific

043317-500MG 043317-1G 043317-5G
101860-200EA 120 USD
101860-200 101860-202 101860-204
3-(2-Chlorophenyl)-1H-pyrazole ≥95%
3-(2-Chlorophenyl)-1H-pyrazole

Hazard: Irritant

Formula: C₉H₇ClN₂
Melting Pt: 76…77 °C
MDL Number: MFCD00111882
CAS Number: 59843-55-9

Order Now

Specification Test Results

Melting point 76-77°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR