N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide ≥95%

Supplier: Matrix Scientific

043316-500MG 043316-1G 043316-5G
101860-194EA 278.46 USD
101860-194 101860-196 101860-198
N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide ≥95%
N-(2,6-Difluorophenyl)-1-hydrazinecarboxamide

Hazard: Irritant

Formula: C₇H₇F₂N₃O
Melting Pt: 161…163 °C
MDL Number: MFCD03791164
CAS Number: 171277-87-5

Order Now

Specification Test Results

Melting point 161-163°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR