6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one

043312-500MG 043312-1G
101860-172EA 278.46 USD
101860-172 101860-174
6-Methyl-4-trifluoromethyl-2(1H)-pyridone ≥95%
6-Methyl-4-trifluoromethyl-2(1H)-pyridone

Hazard: Irritant

Formula: C₇H₆F₃NO
MW: 177.126 g/mol
Melting Pt: 133…135 °C
MDL Number: MFCD00042479
CAS Number: 22123-19-9

Order Now

Specification Test Results

Melting point 133-135°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR