N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide ≥95%

Supplier: Matrix Scientific

043311-500MG 043311-1G 043311-5G
101860-166EA 556.92 USD
101860-166 101860-168 101860-170
N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide ≥95%
N'-Hydroxy-2-[3-(trifluoromethyl)phenoxy]ethanimidamide

Hazard: Irritant

Formula: C₉H₉F₃N₂O₂
MW: 234.18 g/mol
Melting Pt: 76…78 °C
MDL Number: MFCD02090466
CAS Number: 690632-15-6

Order Now

Specification Test Results

Melting point 76-78°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR