5-Hydroxy-2-(3-methoxystyryl)naphtho[1,2-b]furan-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043309-500MG 043309-1G
101860-162EA 375.38 USD
101860-162 101860-164
5-Hydroxy-2-(3-methoxystyryl)naphtho[1,2-b]furan-3-carbaldehyde ≥95%
5-Hydroxy-2-(3-methoxystyryl)naphtho[1,2-b]furan-3-carbaldehyde

Hazard: Irritant

Formula: C₂₂H₁₆O₄
Melting Pt: 232…234 °C
MDL Number: MFCD03791162
CAS Number: 853097-49-1

Order Now

Specification Test Results

Melting point 232-234°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR