5-Hydroxy-2-(3-methoxystyryl)-1-benzofuran-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

043308-500MG 043308-1G 043308-5G
101860-156EA 375.38 USD
101860-156 101860-158 101860-160
5-Hydroxy-2-(3-methoxystyryl)-1-benzofuran-3-carbaldehyde ≥95%
5-Hydroxy-2-(3-methoxystyryl)-1-benzofuran-3-carbaldehyde

Hazard: Irritant

Formula: C₁₈H₁₄O₄
Melting Pt: 164…167 °C
MDL Number: MFCD03791161
CAS Number: 853097-47-9

Order Now

Specification Test Results

Melting point 164-167°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR