2-(4-Methoxyphenyl)-2-phenylacetic acid ≥95%
Supplier: Matrix Scientific
043307-500MG
043307-1G
101860-152EA
747.69
USD
101860-152
101860-154
2-(4-Methoxyphenyl)-2-phenylacetic acid ≥95%
2-(4-Methoxyphenyl)-2-phenylacetic acid
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ Melting Pt: 104-108°C |
CAS Number:
21749-83-7 |
Specification Test Results
Melting point | 104-108°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...