2-Cyclohexyl-1-oxo-3-phenyl-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043304-500MG 043304-1G
101860-144EA 278.46 USD
101860-144 101860-146
2-Cyclohexyl-1-oxo-3-phenyl-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
2-Cyclohexyl-1-oxo-3-phenyl-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₃NO₃
Melting Pt: 237…240 °C
MDL Number: MFCD03016042
CAS Number: 73349-37-8

Order Now

Specification Test Results

Melting point 237-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR