2-Cyclopentyl-3-(2,4-dichlorophenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
Supplier: Matrix Scientific
043303-500MG
043303-1G
043303-5G
101860-138EA
181.54
USD
101860-138
101860-140
101860-142
2-Cyclopentyl-3-(2,4-dichlorophenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
2-Cyclopentyl-3-(2,4-dichlorophenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid
Hazard: Irritant
Formula:
C₂₁H₁₉Cl₂NO₃ Melting Pt: 213…215 °C |
MDL Number:
MFCD03001198 CAS Number: 400073-92-9 |
Specification Test Results
Melting point | 213-215°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...