6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

043302-500MG 043302-1G 043302-5G
101860-132EA 375.38 USD
101860-132 101860-134 101860-136
6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde ≥95%
6-(4-Methylphenyl)imidazo[2,1-b][1,3]thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀N₂OS
Melting Pt: 167…169 °C
MDL Number: MFCD03425723
CAS Number: 82588-42-9

Order Now

Specification Test Results

Melting point 167-169°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR