Methyl-4-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%

Supplier: Matrix Scientific

043301-500MG 043301-1G 043301-5G
101860-126EA 278.46 USD
101860-126 101860-128 101860-130
Methyl-4-(4-formyl-2-nitrophenoxy)benzenecarboxylate ≥95%
Methyl-4-(4-formyl-2-nitrophenoxy)benzenecarboxylate

Hazard: Irritant

Formula: C₁₅H₁₁NO₆
Melting Pt: 103…105 °C
MDL Number: MFCD03001197
CAS Number: 400073-96-3

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR