2-Cyclopentyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043299-500MG 043299-1G 043299-5G
101860-118EA 181.54 USD
101860-118 101860-120 101860-122
2-Cyclopentyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid ≥95%
2-Cyclopentyl-3-(4-methoxyphenyl)-1-oxo-1,2,3,4-tetrahydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₂₂H₂₃NO₄
Melting Pt: 186…188 °C
MDL Number: MFCD03001196
CAS Number: 400073-93-0

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR