5-(4-Methylpiperazino)-2-nitroaniline ≥95%
Supplier: Matrix Scientific
043297-500MG
043297-1G
043297-5G
101860-106EA
29.23
USD
101860-106
101860-108
101860-110
5-(4-Methylpiperazino)-2-nitroaniline ≥95%
5-(4-Methylpiperazino)-2-nitroaniline
Hazard: Irritant
Formula:
C₁₁H₁₆N₄O₂ Melting Pt: 152…155 °C |
MDL Number:
MFCD00545964 CAS Number: 23491-48-7 |
Specification Test Results
Melting point | 152-155°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...