2-Chloro-7-methoxy-3-quinolinecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-7-methoxyquinoline-3-carbaldehyde

043294-1G 043294-5G
101860-092EA 146.15 USD
101860-092 101860-094
2-Chloro-7-methoxy-3-quinolinecarbaldehyde ≥95%
2-Chloro-7-methoxy-3-quinolinecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₈ClNO₂
MW: 221.64 g/mol
Melting Pt: 196…198 °C
MDL Number: MFCD02227048
CAS Number: 68236-20-4

Order Now

Specification Test Results

Melting point 196-198°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR