2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-{2,2-Dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-yl}oxirane

043291-500MG 043291-1G
101860-076EA 556.92 USD
101860-076 101860-078
2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole ≥95%
2,2-Dimethyl-4-(2-oxiranyl)tetrahydrofuro[3,4-d][1,3]dioxole

Hazard: Irritant

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Melting Pt: 81…82 °C
MDL Number: MFCD03425721
CAS Number: 226709-43-9

Order Now

Specification Test Results

Melting point 81-82°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR