{4-[(2,4-Dichlorobenzyl)oxy]phenyl}methanol ≥95%

Supplier: Matrix Scientific

043280-500MG 043280-1G 043280-5G
101860-030EA 375.38 USD
101860-030 101860-032 101860-034
{4-[(2,4-Dichlorobenzyl)oxy]phenyl}methanol ≥95%
{4-[(2,4-Dichlorobenzyl)oxy]phenyl}methanol

Hazard: Irritant

Formula: C₁₄H₁₂Cl₂O₂
Melting Pt: 111…113 °C
MDL Number: MFCD02091003
CAS Number: 400825-70-9

Order Now

Specification Test Results

Melting point 111-113°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR