1-(Methoxycarbonyl)-4-piperidinecarboxylic acid ≥95%
Supplier: Matrix Scientific
043273-500MG
043273-1G
043273-5G
101860-014EA
181.54
USD
101860-014
101860-016
101860-018
1-(Methoxycarbonyl)-4-piperidinecarboxylic acid ≥95%
1-(Methoxycarbonyl)-4-piperidinecarboxylic acid
Hazard: Irritant
Formula:
C₈H₁₃NO₄ Melting Pt: 97…99 °C |
MDL Number:
MFCD02082494 CAS Number: 197585-42-5 |
Specification Test Results
Melting point | 97-99°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...