Methyl-4,5-dihydronaphtho[1,2-b]thiophene-2-carboxylate ≥95%
Supplier: Matrix Scientific
043270-500MG
043270-1G
043270-5G
101859-996EA
921.54
USD
101859-996
101859-998
101860-000
Methyl-4,5-dihydronaphtho[1,2-b]thiophene-2-carboxylate ≥95%
Methyl-4,5-dihydronaphtho[1,2-b]thiophene-2-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₂O₂S Melting Pt: 85…88 °C |
MDL Number:
MFCD01569124 CAS Number: 320423-96-9 |
Specification Test Results
Melting point | 85-88°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...