1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-3,4-dihydronaphthalene-2-carbaldehyde

043269-500MG 043269-1G 043269-5G
101859-990EA 304.62 USD
101859-990 101859-992 101859-994
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde ≥95%
1-Chloro-3,4-dihydro-2-naphthalenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₉ClO
MW: 192.65 g/mol
Boiling Pt: 145…153 °C (5 Torr)
Melting Pt: 36…39 °C
MDL Number: MFCD02257735
CAS Number: 3262-03-1

Order Now

Specification Test Results

Melting point 36-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR