Methyl-4-(3-bromopropoxy)benzenecarboxylate ≥95%
Supplier: Matrix Scientific
043265-500MG
043265-1G
043265-5G
101859-974EA
278.46
USD
101859-974
101859-976
101859-978
Methyl-4-(3-bromopropoxy)benzenecarboxylate ≥95%
Methyl-4-(3-bromopropoxy)benzenecarboxylate
Hazard: Irritant
Formula:
C₁₁H₁₃BrO₃ MW: 273.13 g/mol Melting Pt: 71…73 °C |
MDL Number:
MFCD01814575 CAS Number: 135998-88-8 |
Specification Test Results
Melting point | 71-73°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...