4-(2-Hydroxyethoxy)benzenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(2-Hydroxyethoxy)benzoic acid, 4-(2-Hydroxy-ethoxy)-benzoic acid
043264-500MG
043264-1G
043264-5G
101859-968EA
93.85
USD
101859-968
101859-970
101859-972
4-(2-Hydroxyethoxy)benzenecarboxylic acid ≥95%
4-(2-Hydroxyethoxy)benzenecarboxylic acid
Hazard: Irritant
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Melting Pt: 177…179 °C |
MDL Number:
MFCD00020367 CAS Number: 1711-24-6 |
Specification Test Results
Melting point | 177-179°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...