3-[2-(4-Methylphenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
043256-500MG
043256-1G
043256-5G
101859-922EA
556.92
USD
101859-922
101859-924
101859-926
3-[2-(4-Methylphenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Methylphenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₃ Melting Pt: 213…215 °C |
MDL Number:
MFCD01568766 CAS Number: 1164477-18-2 |
Specification Test Results
Melting point | 213-215°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...