3-[2-(4-Chlorophenoxy)phenyl]acrylic acid ≥95%

Supplier: Matrix Scientific

043255-500MG 043255-1G 043255-5G
101859-916EA 375.38 USD
101859-916 101859-918 101859-920
3-[2-(4-Chlorophenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Chlorophenoxy)phenyl]acrylic acid

Hazard: Irritant

Formula: C₁₅H₁₁ClO₃
Melting Pt: 212…214 °C
MDL Number: MFCD01568765
CAS Number: 320423-53-8

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR