2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-tert-Butylphenoxy)benzaldehyde

043254-500MG 043254-1G
101859-912EA 300 USD
101859-912 101859-914
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde ≥95%
2-[4-(tert-Butyl)phenoxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₇H₁₈O₂
MW: 254.33 g/mol
Melting Pt: 46…48 °C
MDL Number: MFCD01568764
CAS Number: 181297-82-5

Order Now

Specification Test Results

Melting point 46-48°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR