3-[2-(4-Methoxyphenoxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
043253-500MG
043253-1G
043253-5G
101859-906EA
556.92
USD
101859-906
101859-908
101859-910
3-[2-(4-Methoxyphenoxy)phenyl]acrylic acid ≥95%
3-[2-(4-Methoxyphenoxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₆H₁₄O₄ Melting Pt: 208…210 °C |
MDL Number:
MFCD01568763 CAS Number: 1164549-12-5 |
Specification Test Results
Melting point | 208-210°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...