2-(4-Methoxyphenoxy)benzenecarbaldehyde oxime ≥95%
Supplier: Matrix Scientific
043251-500MG
043251-1G
101859-896EA
1109.23
USD
101859-896
101859-898
2-(4-Methoxyphenoxy)benzenecarbaldehyde oxime ≥95%
2-(4-Methoxyphenoxy)benzenecarbaldehyde oxime
Hazard: Irritant
Formula:
C₁₄H₁₃NO₃ Melting Pt: 120…122 °C |
MDL Number:
MFCD01871438 CAS Number: 449778-55-6 |
Specification Test Results
Melting point | 120-122°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...