2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde ≥95%

Supplier: Matrix Scientific

043250-500MG 043250-1G
101859-892EA 1296.92 USD
101859-892 101859-894
2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde ≥95%
2-(4-Methylpiperazino)-1,3-thiazole-5-carbaldehyde

Hazard: Irritant

Formula: C₉H₁₃N₃OS
Melting Pt: 73…75 °C
MDL Number: MFCD01568852
CAS Number: 320423-50-5

Order Now

Specification Test Results

Melting point 73-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR