3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%

Supplier: Matrix Scientific

043249-500MG 043249-1G 043249-5G
101859-886EA 181.54 USD
101859-886 101859-888 101859-890
3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde ≥95%
3-[(2-Chloro-1,3-thiazol-5-yl)methoxy]benzenecarbaldehyde

Hazard: Irritant

Formula: C₁₁H₈ClNO₂S
Melting Pt: 40…42 °C
MDL Number: MFCD03425720
CAS Number: 439094-98-1

Order Now

Specification Test Results

Melting point 40-42°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR